Human recombinant PKC epsilon (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Protein Kinase C Epsilon Type
- UniProtKB:Q02156 (Human)
- Protein/peptide name:PKC epsilon
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MQELEYGPSVDWWALGVLMYEMMAGQPPFEADNEDDLFESILHDDVLYPVWLSKEAVSILKAFMT KNPHKRLGCVASQNGEDAIKQHPFFKEIDWVLLEQKKIKPPFKPRIKTKRDVNNFDQDFTREEPV LTLVDEAIVKQINQEEFKGFSYFGEDLMPLEHHHHHH
- Formulation:Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM DTT, pH 7.4.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together