Entrada de pedido
Spain
ContactUsLinkComponent
 
Human recombinant PITPNA (from E. coli)
 :  Biorbyt
 
Human recombinant PITPNA (from E. coli)
 :  Biorbyt

 

  • Conjugado:
    Unconjugated
  • Tipo de proteína / péptido:
    Recombinant
  • Fuente:
    E. coli
  • Especies:
    Human
  • Condiciones de almacenamiento:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Sinónimos de proteínas:
    Phosphatidylinositol Transfer Alpha Isoform
  • UniProtKB:
    Q00169 (Human)
  • Nombre de proteína / péptido:
    PITPNA
  • Pureza:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MGSSHHHHHHSSGLVPRGSHMVLLKEYRVILPVSVDEYQVGQLYSVAEASKNETGGGEGVEVLVN EPYEKDGEKGQYTHKIYHLQSKVPTFVRMLAPEGALNIHEKAWNAYPYCRTVITNEYMKEDFLIK IETWHKPDLGTQENVHKLEPEAWKHVEAVYIDIADRSQVLSKDYKAEEDPAKFKSIKTGRGPLGP NWKQELVNQKDCPYMCAYKLVTVKFKWWGLQNKVENFIHKQERRLFTNFHRQLFCWLDKWVDLTM DDIRRMEEETKRQLDEMRQKDPVKGMTADD
  • Formulación:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 1mM EDTA, 1mM DTT, pH 8.0.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together