Order Entry
Spain
ContactUsLinkComponent
Human Recombinant CCL27 (from E. coli)
Human Recombinant CCL27 (from E. coli)
Proveedor:  Biorbyt
Certificados
Human Recombinant CCL27 (from E. coli)
Proveedor:  Biorbyt

Especificaciones

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    CC Motif Chemokine 27
  • UniProtKB:
    Q9Y4X3 (Human)
  • Protein/peptide name:
    CCL27
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    FLLPPSTACCTQLYRKPLSDKLLRKVIQVELQEADGDCHLQAFVLHLAQRSICIHPQNPSLSQWF EHQERKLHGTLPKLNFGMLRKMG
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 500mM NaCl, pH 7.5.
  • Tested applications:
    SDS-PAGE

Especificaciones

Información relacionada

Frequently Bought Together