alpha-2-HS-Glycoprotein (from Plasma)
Catalog # BIRBORB82115-1
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:1 mg
- Source:Plasma
- Protein synonyms:HS ELISA Kit
- Protein/peptide name:alpha-2-HS-Glycoprotein
- Purity:Purified.
- Formulation:Lyophilised.