Order Entry
Spain
ContactUsLinkComponent
Human PTH (7-34) (from E. coli)
Human PTH (7-34) (from E. coli)
Supplier:  Biorbyt
Certificates
Human PTH (7-34) (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Biological activity:
    The activity calculated by UMR106 cell/cAMP method corresponding to specific activity of 10,000 Units/mg.
  • Protein synonyms:
    Parathyrin|PTH1 receptor|PTHY_protein.|PTH1|Parathyroid hormone 1|Parathormone|hPTH|PTHR1|PTHR|PTH|Parathyroid hormone|PTH1R
  • Protein/peptide name:
    PTH (7-34)
  • Purity:
    > 97.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized PTH in sterile H2O not less than 100 ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    Leu-Met-His-Asn-Leu-Gly-Lys-His-Leu-Asn-Ser-Met-Glu-Arg-Val-Glu-Trp-Leu-Arg-Lys-Lys-Leu-Gln-Asp-Val-His-Asn-Phe.
  • Formulation:
    The protein (1 mg/ml) was lyophilized after extensive dialyses against 20 mM PBS pH-7.0 and 4% mannitol.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together