Order Entry
Spain
ContactUsLinkComponent
Human HMGB1 (from E. coli)
Human HMGB1 (from E. coli)
Supplier:  Biorbyt
Certificates
Human HMGB1 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Protein synonyms:
    Amphoterin
  • Protein/peptide name:
    HMGB1
  • Purity:
    > 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized HMGB1 in sterile H2O not less than 100 ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    MGKGDPKKPR GKMSSYAFFV QTCREEHKKK HPDASVNFSE FSKKCSERWK TMSAKEKGKF EDMAKADKAR YEREMKTYIP PKGETKKKFK DPNAPKRPPS AFFLFCSEYR PKIKGEHPGL SIGDVAKKLG EMWNNTAADD KQPYEKKAAK LKEKYEKDIA AYRAKGKPDA AKKGVVKAEK SKKKEEEEDE EDEEDEEEEE DEEDEDEEED DDDELEHHHH HH.
  • Formulation:
    The HMG1 (1 mg/ml) was lyophilized after extensive dialyses against 1x PBS pH-7.4.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together