Capillary columns, Rtx®-65TG (fused silica)
Supplier: Restek
Certificates
About this item
High polarity phase columns, Crossbond® diphenyl dimethyl polysiloxane, application specific and specially tested for triglycerides.
- Stable to 370 °C
- Resolves triglycerides by degree of unsaturation as well as by carbon number
Frequently Bought Together