Order Entry
Spain
ContactUsLinkComponent
General purpose gloves, AlphaTec® 37-200 / 37-210
General purpose gloves, AlphaTec® 37-200 / 37-210
Supplier:  Ansell
Certificates
undefined
General purpose gloves, AlphaTec® 37-200 / 37-210
Supplier:  Ansell
Specifications
  • Catalog No:
  • ANSE37-200/9
  • ANSE37-210/8
  • ANSE37-200/12
  • ANSE37-200/7
  • ANSE37-200/11
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finish:
  • Raised large diamond
  • Raised large diamond
  • Raised large diamond
  • Raised large diamond
  • Raised large diamond
  • Cuff style:
  • Gauntlet
  • Gauntlet
  • Gauntlet
  • Gauntlet
  • Gauntlet
  • EN norm:
  • EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5 VIRUS, EN 388:2016 (2001X)
  • EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5 VIRUS, EN 388:2016 (2001X)
  • EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5 VIRUS, EN 388:2016 (2001X)
  • EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5 VIRUS, EN 388:2016 (2001X)
  • EN ISO 374-1:2016 Type B (JKT), EN ISO 374-5 VIRUS, EN 388:2016 (2001X)
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +