GC columns, Rt®-βDEXsp (fused silica)
Supplier: Restek
Certificates
About this item
The Rt®-βDEXsp columns are useful in dual-column configurations, with the Rt-βDEXsm column, for complex enantiomeric separations.
Frequently Bought Together
The Rt®-βDEXsp columns are useful in dual-column configurations, with the Rt-βDEXsm column, for complex enantiomeric separations.