Order Entry
Spain
ContactUsLinkComponent
Anti-C4A Mouse Monoclonal Antibody (CF488A) [clone: TDM16]
Anti-C4A Mouse Monoclonal Antibody (CF488A) [clone: TDM16]
Supplier:  Biotium
Certificates
Anti-C4A Mouse Monoclonal Antibody (CF488A) [clone: TDM16]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC881232-500
  • BNC881232-100
  • Antigen symbol:
  • C4A
  • C4A
  • Antigen name:
  • Complement C4d
  • Complement C4d
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone TDM16
  • Clone TDM16
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 534847 , 720022
  • 534847 , 720022
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together