Order Entry
Spain
ContactUsLinkComponent
 
Anti-ICAM1 Mouse Monoclonal Antibody (CF640R) [clone: 1H4 or Wehi-CAM-1 or W-CAM-1]
 :  Biotium
 
Anti-ICAM1 Mouse Monoclonal Antibody (CF640R) [clone: 1H4 or Wehi-CAM-1 or W-CAM-1]
 :  Biotium

 

  • Catalog No:
  • BNC400988-500
  • BNC400988-100
  • Antigen symbol:
  • ICAM1
  • ICAM1
  • Antigen name:
  • CD54
  • CD54
  • Antigen synonyms:
  • Human rhinovirus receptor|ICAM-1|Cell surface glycoprotein P3.58|Ly 47|Intercellular adhesion molecule 1|CD54|MyD10|Surface antigen of activated B cells|Major group rhinovirus receptor|MALA2
  • Human rhinovirus receptor|ICAM-1|Cell surface glycoprotein P3.58|Ly 47|Intercellular adhesion molecule 1|CD54|MyD10|Surface antigen of activated B cells|Major group rhinovirus receptor|MALA2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone 1H4 or Wehi-CAM-1 or W-CAM-1
  • Clone 1H4 or Wehi-CAM-1 or W-CAM-1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 643447
  • 643447
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together