Order Entry
Spain
ContactUsLinkComponent
Anti-GLG1 Mouse Monoclonal Antibody (CF640R) [clone: GLG1/970]
Anti-GLG1 Mouse Monoclonal Antibody (CF640R) [clone: GLG1/970]
Proveedor:  Biotium
Certificados
Anti-GLG1 Mouse Monoclonal Antibody (CF640R) [clone: GLG1/970]
Proveedor:  Biotium

Especificaciones

  • Catalog No:
  • BTIUBNC400970-500
  • BTIUBNC400970-100
  • Antigen symbol:
  • GLG1
  • GLG1
  • Antigen name:
  • Golgi Glycoprotein 1
  • Golgi Glycoprotein 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone GLG1/970
  • Clone GLG1/970
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 109731
  • 109731
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Especificaciones

Frequently Bought Together