Order Entry
Spain
ContactUsLinkComponent
Anti-LIMP2 Rabbit Polyclonal Antibody
Anti-LIMP2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-LIMP2 Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB400-129SS
  • NOVUNB400-129
  • Antigen symbol:
  • LIMP2
  • LIMP2
  • Antigen name:
  • Lysosome membrane protein 2
  • Lysosome membrane protein 2
  • Antigen synonyms:
  • CD36L2|AMRF|scavenger receptor class B|member 2|LIMPII|85 kDa lysosomal sialoglycoprotein scavenger receptor class B|CD36 antigen (collagen type I receptor|thrombospondin receptor)-like 2 (lysosomal integral membrane protein II)|SCARB2|lysosomal integral membrane protein II|HLGP85
  • CD36L2|AMRF|scavenger receptor class B|member 2|LIMPII|85 kDa lysosomal sialoglycoprotein scavenger receptor class B|CD36 antigen (collagen type I receptor|thrombospondin receptor)-like 2 (lysosomal integral membrane protein II)|SCARB2|lysosomal integral membrane protein II|HLGP85
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 950
  • 950
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together