Order Entry
Spain
ContactUsLinkComponent
Anti-CDKN1C Mouse Monoclonal Antibody (CF647) [clone: KP10]
Anti-CDKN1C Mouse Monoclonal Antibody (CF647) [clone: KP10]
Supplier:  Biotium
Certificates
Anti-CDKN1C Mouse Monoclonal Antibody (CF647) [clone: KP10]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC470879-500
  • BNC470879-100
  • Antigen symbol:
  • CDKN1C
  • CDKN1C
  • Antigen name:
  • Cyclin-dependent kinase inhibitor 1C
  • Cyclin-dependent kinase inhibitor 1C
  • Antigen synonyms:
  • BWCR|p57|Cyclin dependent kinase inhibitor 1C (CDKN1C)|Beckwith Wiedemann syndrome (WBS)|p57Kip2|KIP2|Cyclin dependent kinase inhibitor p57
  • BWCR|p57|Cyclin dependent kinase inhibitor 1C (CDKN1C)|Beckwith Wiedemann syndrome (WBS)|p57Kip2|KIP2|Cyclin dependent kinase inhibitor p57
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone KP10
  • Clone KP10
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 106070
  • 106070
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together