Order Entry
Spain
ContactUsLinkComponent
Anti-GPC3 Mouse Monoclonal Antibody (CF640R) [clone: GPC3/863]
Anti-GPC3 Mouse Monoclonal Antibody (CF640R) [clone: GPC3/863]
Supplier:  Biotium
Certificates
Anti-GPC3 Mouse Monoclonal Antibody (CF640R) [clone: GPC3/863]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC400863-500
  • BNC400863-100
  • Antigen symbol:
  • GPC3
  • GPC3
  • Antigen name:
  • Glypican 3
  • Glypican 3
  • Antigen synonyms:
  • GPC3|Heparan sulphate proteoglycan|SDYS|MXR7|SGBS1|OCI-5|Secreted glypican-3|GTR2-2|Glypican proteoglycan 3|DGSX|Intestinal protein OCI-5
  • GPC3|Heparan sulphate proteoglycan|SDYS|MXR7|SGBS1|OCI-5|Secreted glypican-3|GTR2-2|Glypican proteoglycan 3|DGSX|Intestinal protein OCI-5
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone GPC3/863
  • Clone GPC3/863
  • Reactivity:
  • Human, Rat
  • Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 644108
  • 644108
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together