Anti-CHGC Rabbit Polyclonal Antibody
BOSSBS-12120R
: Bioss
- Antibody type:Primary
- Antigen name:Chromogranin C
- Antigen symbol:CHGC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Chromogranin-C|MEN1|SN.|Secretoneurin included|SgII|Secretogranin-2|Secretogranin II|Secretoneurin|CHGC|SCG2_HUMAN|SCG2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A
- Size:100 μg
- Pk:100 µl
Type: Primary
Antigen: CHGC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat