About this item
This MAb is specific to heavy chain of IgA and shows minimal cross-reaction with heavy chains of other immunoglobulins. It is reactive with all subclasses of Alpha heavy chain. Immunoglobulins are four-chain, Y-shaped, monomeric structures comprised of two identical heavy chains and two identical light chains held together through inter-chain disulfide bonds. The chains form two domains, the Fab (antigen binding) fragment and the Fc (constant) fragment. Immunoglobulin A (IgA) is the main protein of the mucosal immune system. It is generated by B-cells in gut-associated lymphoid tissues. Daily production of IgA exceeds that of any of the other immunoglobulins.IgA exists mainly in dimers but can also exist as polymers or as monomers. Dimers and polymers contain a joining (J) chain that can be bound by the polymeric immunoglobulin receptor (pIgR) for transportation of the molecule to mucosal surfaces. The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Type: Primary
Antigen: IGHA1 & IGHA2
Clonality: Monoclonal
Clone: IA761
Conjugation: CF543
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- BNC430761-500
- BNC430761-100
- Antigen symbol:
- IGHA1 & IGHA2
- IGHA1 & IGHA2
- Antigen name:
- IgA Immunoglobulin
- IgA Immunoglobulin
- Antigen synonyms:
- Ig alpha 1 Chain C Region|Immunoglobulin Heavy Constant Alpha 2|IGHA2|Ig alpha 2 Chain C Region|Immunoglobulin Heavy Constant Alpha 1|Immunoglobulin Am1|IGHA1|Immunoglobulin Am2|A2m Marker
- Ig alpha 1 Chain C Region|Immunoglobulin Heavy Constant Alpha 2|IGHA2|Ig alpha 2 Chain C Region|Immunoglobulin Heavy Constant Alpha 1|Immunoglobulin Am1|IGHA1|Immunoglobulin Am2|A2m Marker
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 543
- CF® 543
- Clone:
- Clone IA761
- Clone IA761
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 699841
- 699841
- Isotype:
- IgG1
- IgG1
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide