Order Entry
Spain
ContactUsLinkComponent
 
Anti-IGHA1 & IGHA2 Mouse Monoclonal Antibody (CF405S) [clone: IA761]
 :  Biotium
 
Anti-IGHA1 & IGHA2 Mouse Monoclonal Antibody (CF405S) [clone: IA761]
 :  Biotium

 

  • Catalog No:
  • BNC040761-500
  • BNC040761-100
  • Antigen symbol:
  • IGHA1 & IGHA2
  • IGHA1 & IGHA2
  • Antigen name:
  • IgA Immunoglobulin
  • IgA Immunoglobulin
  • Antigen synonyms:
  • Ig alpha 1 Chain C Region|Immunoglobulin Heavy Constant Alpha 2|IGHA2|Ig alpha 2 Chain C Region|Immunoglobulin Heavy Constant Alpha 1|Immunoglobulin Am1|IGHA1|Immunoglobulin Am2|A2m Marker
  • Ig alpha 1 Chain C Region|Immunoglobulin Heavy Constant Alpha 2|IGHA2|Ig alpha 2 Chain C Region|Immunoglobulin Heavy Constant Alpha 1|Immunoglobulin Am1|IGHA1|Immunoglobulin Am2|A2m Marker
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 405S
  • CF® 405S
  • Clone:
  • Clone IA761
  • Clone IA761
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 699841
  • 699841
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together