Anti-RIMS1/ABCA4 Rabbit Polyclonal Antibody
Catalog # BOSSBS-11356R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Regulating Synaptic Membrane Exocytosis Protein 1 And ATP Binding Cassette Subfamily A member 4
- Antigen symbol:RIMS1/ABCA4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Regulating synaptic membrane exocytosis protein 1|RIM 1|MGC176677|Serg1 gene name|Rab3-interacting molecule 1|Rims1 gene name|RIM|RIMS1|Rab-3-interacting protein 2|RAB3IP2|MGC167823|Rab3ip1|Regulating synaptic membrane exocytosis 1|ABCA4|CORD7|Rab-3-interacting molecule 1|RIMS1_HUMAN|KIAA0340|Nbla00761.|RAB3 interacting protein 2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: RIMS1/ABCA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat