Specifications
- Antibody type:Primary
- Antigen name:Maternal Embryonic Leucine Zipper Kinase
- Antigen symbol:MELK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:Liquid
- Gene ID:9833
- Antigen synonyms:hMELK|Protein kinase PK38|KIAA0175|Maternal embryonic leucine zipper kinase|MELK|hPK38
- Storage buffer:Aqueous buffered solution containing 100 μg/ml BSA, 50% glycerol and 0,09% sodium azide. Store at −20 °C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4 °C
- Immunogen:130-180/651
- Purification:Purified by Protein A
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Serine/threonine-protein kinase involved in various processes such as cell cycle regulation, self-renewal of stem cells, apoptosis and splicing regulation. Has a broad substrate specificity; phosphorylates BCL2L14, CDC25B, MAP3K5/ASK1 and ZNF622. Acts as an activator of apoptosis by phosphorylating and activating MAP3K5/ASK1. Acts as a regulator of cell cycle, notably by mediating phosphorylation of CDC25B, promoting localization of CDC25B to the centrosome and the spindle poles during mitosis. Plays a key role in cell proliferation and carcinogenesis. Required for proliferation of embryonic and postnatal multipotent neural progenitors. Phosphorylates and inhibits BCL2L14, possibly leading to affect mammary carcinogenesis by mediating inhibition of the pro-apoptotic function of BCL2L14. Also involved in the inhibition of spliceosome assembly during mitosis by phosphorylating ZNF622, thereby contributing to its redirection to the nucleus. May also play a role in primitive hematopoiesis.
Recommended Dilutions: IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.
Type: Primary
Antigen: MELK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat