Order Entry
Spain
ContactUsLinkComponent
Anti-SUMO2 & SUMO3 Mouse Monoclonal Antibody (CF647) [clone: SM23/496]
Anti-SUMO2 & SUMO3 Mouse Monoclonal Antibody (CF647) [clone: SM23/496]
Supplier:  Biotium
Certificates
Anti-SUMO2 & SUMO3 Mouse Monoclonal Antibody (CF647) [clone: SM23/496]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC470496-100
  • BNC470496-500
  • Antigen symbol:
  • SUMO2 + SUMO3
  • SUMO2 + SUMO3
  • Antigen name:
  • Small ubiquitin-related modifier 2 and 3
  • Small ubiquitin-related modifier 2 and 3
  • Antigen synonyms:
  • Recombinant human SUMO2 protein
  • Recombinant human SUMO2 protein
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 647
  • CF® 647
  • Clone:
  • Clone SM23/496
  • Clone SM23/496
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together