Order Entry
Spain
ContactUsLinkComponent
 
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF488A) [clone: L243]
 :  Biotium
 
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF488A) [clone: L243]
 :  Biotium

 

  • Catalog No:
  • BNC880458-500
  • BNC880458-100
  • Antigen symbol:
  • HLA-DRB1
  • HLA-DRB1
  • Antigen name:
  • HLA-DRB
  • HLA-DRB
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone L243
  • Clone L243
  • Reactivity:
  • Baboon, Monkey, Squirrel, Chimpanzee, Human, Dog
  • Baboon, Monkey, Squirrel, Chimpanzee, Human, Dog
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 534322
  • 534322
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together