Order Entry
Spain
ContactUsLinkComponent
Anti-TP53 Rabbit Monoclonal Antibody (CF488A) [clone: PAb122]
Anti-TP53 Rabbit Monoclonal Antibody (CF488A) [clone: PAb122]
Supplier:  Biotium
Certificates
Anti-TP53 Rabbit Monoclonal Antibody (CF488A) [clone: PAb122]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC880338-100
  • BNC880338-500
  • Antigen symbol:
  • TP53
  • TP53
  • Antigen name:
  • p53
  • p53
  • Antigen synonyms:
  • Transformation Related Protein 53 (TRP53)|Tumor Protein p53|BCC7|Cellular Tumor Antigen p53|Tumor Suppressor p53|TP53|LFS1|Antigen NY-CO-13
  • Transformation Related Protein 53 (TRP53)|Tumor Protein p53|BCC7|Cellular Tumor Antigen p53|Tumor Suppressor p53|TP53|LFS1|Antigen NY-CO-13
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone PAb122
  • Clone PAb122
  • Reactivity:
  • Hamster, Monkey, Human, Dog, Rat, Mouse
  • Hamster, Monkey, Human, Dog, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 654481
  • 654481
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together