Order Entry
Spain
ContactUsLinkComponent
 
Anti-HIF1A Mouse Monoclonal Antibody (CF640R) [clone: HIF1A-84]
 :  Biotium
 
Anti-HIF1A Mouse Monoclonal Antibody (CF640R) [clone: HIF1A-84]
 :  Biotium

 

  • Catalog No:
  • BNC400084-100
  • BNC400084-500
  • Antigen symbol:
  • HIF1A
  • HIF1A
  • Antigen name:
  • HIF1-alpha
  • HIF1-alpha
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone HIF1A-84
  • Clone HIF1A-84
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 597216
  • 597216
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together