Order Entry
Spain
ContactUsLinkComponent
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF640R) [clone: LN-3]
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF640R) [clone: LN-3]
Supplier:  Biotium
Certificates
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF640R) [clone: LN-3]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC400057-100
  • BNC400057-500
  • Antigen symbol:
  • HLA-DRB1
  • HLA-DRB1
  • Antigen name:
  • HLA-DRB
  • HLA-DRB
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 640R
  • CF® 640R
  • Clone:
  • Clone LN-3
  • Clone LN-3
  • Reactivity:
  • Monkey, Human
  • Monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 534322
  • 534322
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together