Order Entry
Spain
ContactUsLinkComponent
Anti-S100A8 & S100A9 Mouse Monoclonal Antibody (CF568) [clone: MAC387]
Anti-S100A8 & S100A9 Mouse Monoclonal Antibody (CF568) [clone: MAC387]
Supplier:  Biotium
Certificates
Anti-S100A8 & S100A9 Mouse Monoclonal Antibody (CF568) [clone: MAC387]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC680035-500
  • BNC680035-100
  • Antigen symbol:
  • S100A8 & S100A9
  • S100A8 & S100A9
  • Antigen name:
  • Calgranulin B / MRP-8/-14 / MAC 387 / S100A8/A9 / Macrophage Ag
  • Calgranulin B / MRP-8/-14 / MAC 387 / S100A8/A9 / Macrophage Ag
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone MAC387
  • Clone MAC387
  • Reactivity:
  • Mouse, Cat, Baboon, Pig, Rabbit, Horse, Monkey, Goat, Human, Bovine, Guinea pig, Dog, Rat
  • Mouse, Cat, Baboon, Pig, Rabbit, Horse, Monkey, Goat, Human, Bovine, Guinea pig, Dog, Rat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 416073 (S100A8/Calgranulin A/MRP-8) , 112405 (S100A9/Calgranulin B/MRP-14)
  • 416073 (S100A8/Calgranulin A/MRP-8) , 112405 (S100A9/Calgranulin B/MRP-14)
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together