Dimethyl sulphoxide ≥99%, liquid
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:67-68-5
:
Formula:(CH₃)₂SO
MDL Number:MFCD00002089
:78.14 g/mol
Boiling Pt:189 °C (1013 hPa) (lit.)
Melting Pt:16…19 °C (lit.)
Density:1.104 g/cm³ (20 °C) (lit.)
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:>99%
Specification Test Results
Frequently Bought Together