Sodium dihydrogen phosphate, anhydrous ≥98%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7558-80-7
:
Formula:NaH₂PO₄
MDL Number:MFCD00003527
:119.98 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test :Passes
- Purity:≥98%
- Moisture Content:≤1%
- pH (1% aq soln):3 - 5
Specification Test Results
Frequently Bought Together