Glycerine, liquid
Supplier: MP Biomedicals
Jump to:
About this item
Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
- CAS Number:56-81-5
- Formula:HOCH₂CH(OH)CH₂OH
- MDL Number:MFCD00004722
- Molecular Weight:92.09 g/mol
2 Options Available Below
Product Details & Documents
Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
- CAS Number:56-81-5
- Formula:HOCH₂CH(OH)CH₂OH
- MDL Number:MFCD00004722
- Molecular Weight:92.09 g/mol
Product Family Options
Product Information
- PkAvailabilityPrice



