Order Entry
Spain
ContactUsLinkComponent
 
Sodium L(+)-ascorbate ≥99%, white powder
 
undefined
Sodium L(+)-ascorbate ≥99%, white powder

 

  • Identity Test:
    Passes
  • Purity:
    ≥99%
  • FTIR:
    Conforms to Standard
  • Optical Rotation (c=1, water):
    +100° to +110°

 

Frequently Bought Together