DMP (Dimethyl pimelimidate dihydrochloride), white crystalline powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:58537-94-3
:
Formula:C₉H₁₈N₂O₂
MDL Number:MFCD00038942
:259.17 g/mol
Melting Pt:122 °C (dec., lit.)
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
Specification Test Results
Frequently Bought Together