Anti-PSMA3 Rabbit Polyclonal Antibody
BSBTPB10089
- Antibody type:Primary
- Antigen name:Proteasome subunit alpha 3
- Antigen symbol:PSMA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5684
- Antigen synonyms:PSC3|HC8
- Size:100 μg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Proteasome subunit alpha type-3(PSMA3) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: PSMA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat