Anti-GNB1 Rabbit Polyclonal Antibody
Catalog # BSBTPB10067
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Guanine nucleotide binding protein (G protein), beta 1
- Antigen symbol:GNB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2782
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-1(GNB1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Type: Primary
Antigen: GNB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat