Order Entry
Spain
ContactUsLinkComponent
 
Anti-CYP3A4 Rabbit Polyclonal Antibody
  BSBTPB10055
undefined
Anti-CYP3A4 Rabbit Polyclonal Antibody
  BSBTPB10055

 

  • Antibody type:
    Primary
  • Antigen name:
    Cytochrome P450, Family 3, Subfamily A, Polypeptide 4
  • Antigen symbol:
    CYP3A4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    1576
  • Antigen synonyms:
    P450C3|NF-25|CP34|CYP3A|CYPIIIA4|CYP3A3|CYPIIIA3|HLP|P450PCN1|CP33
  • Size:
    100 μg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Cytochrome P450 3A4(CYP3A4) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: CYP3A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human