Anti-PTH Mouse Monoclonal Antibody [clone: 3H9]
Catalog # ABNOMAB13404
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:PTH
- Clonality:Monoclonal
- Clone:3H9
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Gene ID:5741
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of human PTH.
Type: Primary
Antigen: PTH
Clonality: Monoclonal
Clone: 3H9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human