Order Entry
Spain
ContactUsLinkComponent
Anti-HLA-DRB1 Mouse Monoclonal Antibody [clone: LN-3]
Anti-HLA-DRB1 Mouse Monoclonal Antibody [clone: LN-3]
Catalog # ABNOMAB13306
Supplier:  Abnova
Anti-HLA-DRB1 Mouse Monoclonal Antibody [clone: LN-3]
Catalog # ABNOMAB13306
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HLA-DRB1
  • Clonality:
    Monoclonal
  • Clone:
    LN-3
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Gene ID:
    3123
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against native human HLA-DRB1.

Type: Primary
Antigen: HLA-DRB1
Clonality: Monoclonal
Clone: LN-3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human