Order Entry
Spain
ContactUsLinkComponent
 
Anti-KRT7/KRT8 Mouse Monoclonal Antibody [clone: CAM 5.2]
  ILM84911-C1
Anti-KRT7/KRT8 Mouse Monoclonal Antibody [clone: CAM 5.2]
  ILM84911-C1

 

  • Antibody type:
    Primary
  • Antigen name:
    Keratin 7/Keratin 8
  • Antigen symbol:
    KRT7/KRT8
  • Clonality:
    Monoclonal
  • Clone:
    CAM 5.2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Seq:
    03
  • Environmentally Preferable:
  • Form:
    Concentrated
  • Gene ID:
    3855, 3856
  • Antigen synonyms:
    CK7|CYKER K2C8|CYK8|SCL|K2C7|KO|K8|CARD2|K7|CK8
  • Molecular weight:
    53 kDa
  • Storage temperature:
    2°C - 8°C
  • Tested applications:
    Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

The Cytokeratin 7 & 8 antibody has a primary reactivity with human cytokeratin proteins that correspond to Moll's peptide 7 (cytokeratin 7) and Moll's peptide 8 (cytokeratin 8).1-4 These antigens are present on secretory epithelia of normal human tissue, but not on stratified squamous epithelium.1 Its antibody stains most epithelial-derived tissue, including liver, renal tubular epithelium, and hepatocellular and renal cell carcinomas. It may not react with some squamous cell carcinomas. 2, 5, 6

  • Control: Adenocarcinomas, pancreas, prostate, salivary gland
  • Dilution Range: 1:400-1:800 for immunohistochemical staining

References:
1. Moll R, et al, Cell. 1982;31:11-24
2. Makin C,et al, J Clin Pathol. 1984;37:975
3. Cooper D, et al, Lab In vest. 1985;52:243-256
4. Smedts F, et al, Am J Pathol. 1990;136:657-667
5. Battifora H.Progress in Surgical Pathology. Vol 8: Field & Wood Medical Publishers; 1990:1-15
6. Johnson A, et al, Br J Cancer. 1985;52:159-165
7. Wayne, PA: Clinical and Laboratory Standards Institute; 2005. CLSI document M29-A3
8. Centers for Disease Control. MMWR. 1988;37:377-388

Pretreatment: Heat induced epitope retrieval in 10 mM citrate buffer, pH 6,0 for 20 minutes or Protease K treatment for 10 min at 37 °C is required for IHC staining on formalin-fixed, paraffin embedded tissue sections.

Type: Primary
Antigen: KRT7/KRT8
Clonality: Monoclonal
Clone: CAM 5.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human