HPLC columns, ZORBAX® 80 Å Extend-C18
Supplier: Agilent
Certificates
About this item
High sensitivity for LC/MS separations of peptides.
- High efficiency and long life at high pH (up to 11,5)
- Unique bidentate bonding and double encapping provides high pH stability
- Improve retention, resolution and peak shape of basic compounds
More efficiency and better peak shape than polymer-based columns.
Frequently Bought Together