Order Entry
Spain
ContactUsLinkComponent
 
Perchloric acid 60% in water for analysis
  ACRO452850010
 :  
undefined
Perchloric acid 60% in water for analysis
  ACRO452850010
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Appearance:
    Clear colorless Liquid
  • Color scale:
    ≤10 APHA
  • Titration with NaOH:
    60.0 to 65.0 %
  • Chloride (Cl):
    ≤3 ppm
  • Chlorate (ClO3):
    ≤10 ppm
  • Sulfate (SO4):
    ≤10 ppm
  • Silicate (as SiO2):
    ≤5 ppm
  • Cadmium (Cd):
    ≤0.05 ppm
  • Calcium (Ca):
    ≤0.5 ppm
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤1 ppm
  • Lead (Pb):
    ≤0.05 ppm
  • Magnesium (Mg):
    ≤0.5 ppm
  • Nickel (Ni):
    ≤0.1 ppm
  • Nitrogen (N):
    ≤10 ppm
  • Potassium (K):
    ≤0.1 ppm
  • Sodium (Na):
    ≤0.5 ppm
  • Zinc (Zn):
    ≤0.1 ppm

 

 

 :
 :100.46 g/mol
 :203 °C (760 mmHg)
 :–18 °C
 :1.66 g/cm³
 :113 °C
 :Ambient
 :1873
 :5.1,I
 :GHS05
 :GHS03
 :GHS07
 :GHS08
 :7601-90-3
 :HClO₄
 :MFCD00011325