Perchloric acid ∼70% in water, extra pure
Catalog # ACRO451770025
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Appearance:Clear solution
- Color scale:≤10 APHA
- Residue after ignition:≤0.003 %
- Titration with NaOH:70 to 72 %
- Chloride (Cl):≤3 ppm
- Sulfate (SO4):≤5 ppm
- Iron (Fe):≤1 ppm
- Heavy metals (as Pb):≤1 ppm
Specification Test Results
About this item
:
:100.46 g/mol
Boiling Pt:203 °C (760 mmHg)
Melting Pt:–18 °C
Density:1.66 g/cm³
Flash Pt:113 °C
Storage Temperature:Ambient
UN:1873
ADR:5.1,I
:GHS05
:GHS03
:GHS07
:GHS08
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325