Order Entry
Spain
ContactUsLinkComponent
1,4-Dioxane, extra dry 99.8% stabilised, AcroSeal®
Flammable
Irritant
Health
1,4-Dioxane, extra dry 99.8% stabilised, AcroSeal®
Catalog # 450171000.
CAS Number:  
undefined
Flammable
Irritant
Health
1,4-Dioxane, extra dry 99.8% stabilised, AcroSeal®
Catalog # 450171000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.005 %
  • GC:
    ≥99.75 %
  • Stabilizer:
    2 to 5 ppm BHT
  • Residue after evaporation:
    ≤0.001 %
  • Acidity (CH3COOH):
    ≤0.002
  • Peroxides:
    ≤0.005 % (as H2O2)

Specification Test Results

About this item

 :
 :88.11 g/mol
Boiling Pt:101 °C (760 mmHg)
Melting Pt:12 °C
Density:1.034 g/cm³
Flash Pt:12 °C
UN:1165
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:123-91-1
Formula:C₄H₈O₂
MDL Number:MFCD00006571