Gallium(III) chloride, anhydrous 99.99+% (trace metals basis), beads -10 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13450-90-3
:
Formula:Cl₃Ga
MDL Number:MFCD00011018
:176.08 g/mol
Boiling Pt:201 °C (760 mmHg)
Melting Pt:78 °C
Density:2,29 g/cm³ (80 °C)
Storage Temperature:Ambient
UN:3260
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to off-white Beads
- Total trace metal impurities:≤100 ppm
Specification Test Results
Frequently Bought Together