Order Entry
Spain
ContactUsLinkComponent
N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide 97%
N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide 97%
Catalog # ACRO440580050
CAS Number:  
undefined
N-(tert-Butoxycarbonyl)-L-valine N'-methoxy-N'-methylamide 97%
Catalog # ACRO440580050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Low melting solid or liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %
  • Specific optical rotation:
    -15° to -20° (20°C, 589 nm) (c=1 in methanol)

Specification Test Results

About this item

 :
Boiling Pt:248 °C (760 mmHg)
Melting Pt:34…37 °C
Flash Pt:113 °C
 :260.334 g/mol
Storage Temperature:Refrigerator
CAS Number:87694-52-8
Formula:C₁₂H₂₄N₂O₄
MDL Number:MFCD00151897