Order Entry
Spain
ContactUsLinkComponent
 
Polyvinyl alcohol, M.W. ∼ 31,000-50,000 85-89 %, hydrolysed
 
undefined
Polyvinyl alcohol, M.W. ∼ 31,000-50,000 85-89 %, hydrolysed

 

  • Appearance:
    White to cream Granular powder
  • Infrared spectrum:
    Conforms
  • Viscosity:
    5.2 to 6.2 cP (4 % in water)
  • Degree of hydrolysis:
    87.0 to 89.0 mol %
  • Ash:
    ≤0.7 % (as Na2O)
  • pH:
    4.5 to 6.5 (4 % in water)
  • Volatile matter:
    ≤5 %

 

 

Frequently Bought Together