2,2'-Azobis(2-methylpropionamidine)dihydrochloride (AAPH) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2997-92-4
:
Formula:C₈H₁₈N₆·2HCl
MDL Number:MFCD00142725
:271.19 g/mol
Storage Temperature:Refrigerator
UN:3088
ADR:4.2,II
:GHS07
:GHS02
:GHS09
Specification Test Results
- Appearance:White to off-white Granules
- Infrared spectrum:Conforms
- Titration Argentometric:97.5 to 102.5 % (on dry substance)
- Loss on drying:≤1 %
Specification Test Results
Related Information
Frequently Bought Together