Order Entry
Spain
ContactUsLinkComponent
Triphenylphosphine 1.2 - 1.5 mmol/g on resin, cross-linked with 1% DVB 200-400 mesh
Irritant
Triphenylphosphine 1.2 - 1.5 mmol/g on resin, cross-linked with 1% DVB 200-400 mesh
Catalog # ACRO366840050
CAS Number:  
undefined
Irritant
Triphenylphosphine 1.2 - 1.5 mmol/g on resin, cross-linked with 1% DVB 200-400 mesh
Catalog # ACRO366840050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to yellow Beads
  • Particle size:
    200 to 400 mesh
  • Crosslinking:
    1 % DVB
  • Loading:
    1 to 1.5 mmol/g

Specification Test Results

About this item

 :
 :262.29 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:39319-11-4
Formula:C₁₈H₁₅P
MDL Number:MFCD20489348