Order Entry
Spain
ContactUsLinkComponent
Sodium biphenyl 20% (w/w) in diethylene glycol diethyl ether
Corrosive
Flammable
Environment
Sodium biphenyl 20% (w/w) in diethylene glycol diethyl ether
Catalog # ACRO419423000
CAS Number:  
undefined
Corrosive
Flammable
Environment
Sodium biphenyl 20% (w/w) in diethylene glycol diethyl ether
Catalog # ACRO419423000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    300 mL
  • Seq:
    2625
  • Pk Info:
    20 x 15ml

Specifications

Specification Test Results

  • Appearance:
    Dark green to blue-black Solution (May partially crystallize)
  • Titration with HClO4:
    15 to 25 % w/w
  • Refractive index:
    1.5480 to 1.5500 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

About this item

CAS Number:5137-46-2
 :
Formula:C₁₂H₁₀Na
MDL Number:MFCD00013182
 :177.202 g/mol
Boiling Pt:188 °C
Density:1.003 g/cm³
Flash Pt:46 °C
UN:2920
ADR:8,II
 :GHS05
 :GHS02
 :GHS09
Storage Temperature:Refrigerator