Amberlyst® 15, dry ion-exchange resin
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9037-24-5
:39389-20-3
MDL Number:MFCD00145841
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Beads
- Water:≤1.5 %
- Particle size:≤1.0 % (< 0.300 mm)
- Total weight capacity:≥4.70 eq/kg (H)
Specification Test Results
Frequently Bought Together