Order Entry
Spain
ContactUsLinkComponent
Palladium 0.5%, on 1/8-inch alumina pellets
Palladium 0.5%, on 1/8-inch alumina pellets
Certificates
undefined
Palladium 0.5%, on 1/8-inch alumina pellets

Specification Test Results

  • Appearance:
    Grey Pellets
  • Assay:
    0.5 % (Pd)
  • Loss on drying:
    1 % (typically)
  • Additional info:
    typical properties:

Specification Test Results

Related Information

Frequently Bought Together