New methylene Blue N, pure
Catalog # ACRO415530025
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 g
- Seq:3122
Specifications
Specification Test Results
- Appearance:Green to dark brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- Dye content:ca 70%
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:347.91 g/mol
Melting Pt:190 °C
Storage Temperature:Ambient
CAS Number:1934-16-3
Formula:C₁₈H₂₂ClN₃S
MDL Number:MFCD00135878